Assay Details
| Gene Symbol: ste14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274864.1 | NP_001261793.1 | 2 - 3 | 731 | 69 | |
| NM_140366.3 | NP_648623.1 | 2 - 3 | 731 | 69 | ||
| GenBank mRNA | AY070700.1 | - | 2 - 3 | 747 | 69 | |
| AY071003.1 | - | 2 - 3 | 719 | 69 | ||
More Information
Set Membership: |
|

ste14
Genomic Map