Assay Details
| Gene Symbol: eIF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258978.3 | NP_001245907.1 | 2 - 3 | 302 | 60 | |
| NM_001273188.1 | NP_001260117.1 | 2 - 3 | 458 | 60 | ||
| NM_057247.5 | NP_476595.1 | 2 - 3 | 302 | 60 | ||
| NM_164668.3 | NP_723137.1 | 2 - 3 | 302 | 60 | ||
| NM_164669.3 | NP_723138.1 | 2 - 3 | 458 | 60 | ||
| NM_164670.4 | NP_723139.1 | 2 - 3 | 689 | 60 | ||
| GenBank mRNA | AF145621.1 | - | 3405 | 60 | ||
| AY069283.1 | - | 2 - 3 | 297 | 60 | ||
| AY121623.1 | - | 1193 | 60 | |||
| BT099924.1 | - | 2 - 3 | 425 | 60 | ||
More Information
Set Membership: |
|

eIF4A
Genomic Map