Assay Details
| Gene Symbol: eIF4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258978.3 | NP_001245907.1 | 3 - 4 | 914 | 76 | |
| NM_001273188.1 | NP_001260117.1 | 3 - 4 | 1070 | 76 | ||
| NM_057247.5 | NP_476595.1 | 3 - 4 | 914 | 76 | ||
| NM_164668.3 | NP_723137.1 | 3 - 4 | 914 | 76 | ||
| NM_164669.3 | NP_723138.1 | 3 - 4 | 1070 | 76 | ||
| NM_164670.4 | NP_723139.1 | 3 - 4 | 1301 | 76 | ||
| GenBank mRNA | AF145621.1 | - | 4017 | 76 | ||
| AY069283.1 | - | 3 - 4 | 909 | 76 | ||
| AY121623.1 | - | 1805 | 76 | |||
| BT099924.1 | - | 3 - 4 | 1037 | 76 | ||
More Information
Set Membership: |
|

eIF4A
Genomic Map