Assay Details
| Gene Symbol: mmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298744.1 | NP_001285673.1 | 2 - 3 | 643 | 82 | |
| NM_135188.3 | NP_609032.1 | 2 - 3 | 662 | 82 | ||
| NM_164690.2 | NP_723183.1 | 2 - 3 | 500 | 82 | ||
| GenBank mRNA | AF145686.1 | - | 2 - 3 | 647 | 82 | |
More Information
Set Membership: |
|

mmy
Genomic Map