Assay Details
| Gene Symbol: eya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078768.4 | NP_523492.1 | 2 - 3 | 576 | 140 | |
| NM_164693.2 | NP_723188.1 | 2 - 3 | 331 | 140 | ||
| GenBank mRNA | AY047539.1 | - | 2 - 3 | 540 | 140 | |
| L08501.1 | - | 2 - 3 | 497 | 140 | ||
| L08502.1 | - | 2 - 3 | 331 | 140 | ||
More Information
Set Membership: |
|

eya
Genomic Map