Assay Details
| Gene Symbol: eya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258984.1 | NP_001245913.1 | 2 - 3 | 1839 | 73 | |
| NM_078768.4 | NP_523492.1 | 3 - 4 | 2327 | 73 | ||
| NM_164693.2 | NP_723188.1 | 3 - 4 | 2082 | 73 | ||
| GenBank mRNA | AY047539.1 | - | 3 - 4 | 2291 | 73 | |
| L08501.1 | - | 3 - 4 | 2248 | 73 | ||
| L08502.1 | - | 3 - 4 | 2082 | 73 | ||
More Information
Set Membership: |
|

eya
Genomic Map