Assay Details
| Gene Symbol: Liprin-alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273224.1 | NP_001260153.1 | 13 - 14 | 3681 | 81 | |
| NM_001273225.1 | NP_001260154.1 | 14 - 15 | 3826 | 81 | ||
| NM_001273226.1 | NP_001260155.1 | 13 - 14 | 3598 | 81 | ||
| NM_135223.3 | NP_609067.2 | 13 - 14 | 3598 | 81 | ||
| NM_164707.3 | NP_723208.1 | 13 - 14 | 3691 | 81 | ||
| GenBank mRNA | BT004484.1 | - | 13 - 14 | 3562 | 81 | |
More Information
Set Membership: |
|

Liprin-alpha
Genomic Map