Assay Details
| Gene Symbol: Liprin-alpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273224.1 | NP_001260153.1 | 14 - 15 | 3803 | 92 | |
| NM_001273225.1 | NP_001260154.1 | 15 - 16 | 3948 | 92 | ||
| NM_001273226.1 | NP_001260155.1 | 14 - 15 | 3720 | 92 | ||
| NM_135223.3 | NP_609067.2 | 14 - 15 | 3720 | 92 | ||
| NM_164707.3 | NP_723208.1 | 14 - 15 | 3813 | 92 | ||
| GenBank mRNA | BT004484.1 | - | 14 - 15 | 3684 | 92 | |
More Information
Set Membership: |
|

Liprin-alpha
Genomic Map