Assay Details
| Gene Symbol: nrv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014476.2 | NP_001014476.1 | 2 - 3 | 257 | 62 | |
| NM_057821.4 | NP_477169.1 | 2 - 3 | 288 | 62 | ||
| NM_164711.2 | NP_723216.1 | 2 - 3 | 464 | 62 | ||
| GenBank mRNA | BT025048.1 | - | 2 - 3 | 255 | 62 | |
More Information
Set Membership: |
|

nrv2
Genomic Map