Assay Details
| Gene Symbol: nrv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014475.2 | NP_001014475.1 | 5 - 6 | 1173 | 57 | |
| NM_001014476.2 | NP_001014476.1 | 6 - 7 | 979 | 57 | ||
| NM_001273235.1 | NP_001260164.1 | 6 - 7 | 1665 | 57 | ||
| NM_057820.4 | NP_477168.1 | 7 - 8 | 1698 | 57 | ||
| NM_057821.4 | NP_477169.1 | 6 - 7 | 1010 | 57 | ||
| NM_164711.2 | NP_723216.1 | 6 - 7 | 1186 | 57 | ||
| GenBank mRNA | AY060289.1 | - | 5 - 6 | 1160 | 57 | |
| BT025048.1 | - | 6 - 7 | 977 | 57 | ||
More Information
Set Membership: |
|

nrv2
Genomic Map