Assay Details
| Gene Symbol: sens-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298763.1 | NP_001285692.1 | 1 - 2 | 1123 | 65 | |
| NM_164716.2 | NP_723223.1 | 1 - 2 | 740 | 65 | ||
| GenBank mRNA | BT133368.1 | - | 1 - 2 | 740 | 65 | |
More Information
Set Membership: |
|

sens-2
Genomic Map