Assay Details
| Gene Symbol: Mnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042877.2 | NP_001036342.1 | 2 - 3 | 1300 | 63 | |
| NM_078774.4 | NP_523498.2 | 2 - 3 | 1300 | 63 | ||
| NM_164739.3 | NP_723252.2 | 2 - 3 | 556 | 63 | ||
| GenBank mRNA | AY084134.1 | - | 2 - 3 | 551 | 63 | |
| BT003213.1 | - | 2 - 3 | 551 | 63 | ||
More Information
Set Membership: |
|

Mnn1
Genomic Map