Assay Details
| Gene Symbol: Mnn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042877.2 | NP_001036342.1 | 3 - 4 | 1768 | 101 | |
| NM_078774.4 | NP_523498.2 | 3 - 4 | 1768 | 101 | ||
| NM_164739.3 | NP_723252.2 | 3 - 4 | 1024 | 101 | ||
| GenBank mRNA | AY084134.1 | - | 3 - 4 | 1019 | 101 | |
| BT003213.1 | - | 3 - 4 | 1019 | 101 | ||
More Information
Set Membership: |
|

Mnn1
Genomic Map