Assay Details
| Gene Symbol: wg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078778.5 | NP_523502.1 | 3 - 4 | 1105 | 69 | |
| GenBank mRNA | BT133499.1 | - | 3 - 4 | 1103 | 69 | |
| M17230.1 | - | 3 - 4 | 1089 | 69 | ||
More Information
Set Membership: |
|

wg
Genomic Map