Assay Details
| Gene Symbol: ninaC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273260.1 | NP_001260189.1 | 6 - 7 | 2805 | 71 | |
| NM_001273261.1 | NP_001260190.1 | 6 - 7 | 2805 | 71 | ||
| NM_078779.3 | NP_523503.2 | 6 - 7 | 2839 | 71 | ||
| NM_164747.1 | NP_723271.1 | 6 - 7 | 2805 | 71 | ||
| GenBank mRNA | BT150399.1 | - | 6 - 7 | 2823 | 71 | |
More Information
Set Membership: |
|

ninaC
Genomic Map