Assay Details
| Gene Symbol: muc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273267.1 | NP_001260196.1 | 3 - 4 | 933 | 82 | |
| NM_135274.2 | NP_609118.1 | 3 - 4 | 1069 | 82 | ||
| NM_164749.1 | NP_723274.1 | 4 - 5 | 693 | 82 | ||
| GenBank mRNA | BT023873.1 | - | 3 - 4 | 1069 | 82 | |
More Information
Set Membership: |
|

muc
Genomic Map