Assay Details
| Gene Symbol: Akap200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298822.1 | NP_001285751.1 | 2 - 3 | 354 | 65 | |
| NM_001298823.1 | NP_001285752.1 | 2 - 3 | 354 | 65 | ||
| NM_001298824.1 | NP_001285753.1 | 2 - 3 | 403 | 65 | ||
| NM_058111.4 | NP_477459.1 | 1 - 2 | 311 | 65 | ||
| NM_058112.4 | NP_477460.1 | 2 - 3 | 354 | 65 | ||
| NM_164820.2 | NP_723395.1 | 2 - 3 | 403 | 65 | ||
| NM_175984.1 | NP_787998.1 | 2 - 3 | 407 | 65 | ||
| GenBank mRNA | BT010288.1 | - | 2 - 3 | 334 | 65 | |
More Information
Set Membership: |
|

Akap200
Genomic Map