Assay Details
| Gene Symbol: borr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135435.5 | NP_609279.1 | 2 - 3 | 884 | 74 | |
| NM_164850.4 | NP_723450.1 | 2 - 3 | 896 | 74 | ||
| GenBank mRNA | AY061432.1 | - | 2 - 3 | 829 | 74 | |
| BT001609.1 | - | 2 - 3 | 891 | 74 | ||
More Information
Set Membership: |
|

borr
Genomic Map