Assay Details
| Gene Symbol: Eaat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201822.2 | NP_001188751.1 | 5 - 6 | 874 | 71 | |
| NM_001382081.1 | NP_001369122.1 | 5 - 6 | 902 | 71 | ||
| NM_001382082.1 | NP_001369123.1 | 5 - 6 | 902 | 71 | ||
| NM_058080.4 | NP_477428.1 | 5 - 6 | 902 | 71 | ||
| NM_164853.3 | NP_723454.1 | 5 - 6 | 706 | 71 | ||
| NM_164854.3 | NP_723455.1 | 5 - 6 | 768 | 71 | ||
| GenBank mRNA | AF001784.1 | - | 4 - 5 | 493 | 71 | |
| BT044100.1 | - | 5 - 6 | 759 | 71 | ||
| D86739.2 | - | 5 - 6 | 879 | 71 | ||
More Information
Set Membership: |
|

Eaat1
Genomic Map