Assay Details
| Gene Symbol: Eaat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201822.2 | NP_001188751.1 | 7 - 8 | 1098 | 73 | |
| NM_001382081.1 | NP_001369122.1 | 7 - 8 | 1126 | 73 | ||
| NM_001382082.1 | NP_001369123.1 | 7 - 8 | 1126 | 73 | ||
| NM_058080.4 | NP_477428.1 | 7 - 8 | 1126 | 73 | ||
| NM_164853.3 | NP_723454.1 | 7 - 8 | 930 | 73 | ||
| NM_164854.3 | NP_723455.1 | 7 - 8 | 992 | 73 | ||
| GenBank mRNA | BT044100.1 | - | 7 - 8 | 983 | 73 | |
More Information
Set Membership: |
|

Eaat1
Genomic Map