Assay Details
| Gene Symbol: numb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273362.1 | NP_001260291.1 | 4 - 5 | 1147 | 72 | |
| NM_078799.3 | NP_523523.2 | 3 - 4 | 1122 | 72 | ||
| NM_164855.2 | NP_723460.1 | 3 - 4 | 769 | 72 | ||
| GenBank mRNA | BT056305.1 | - | 3 - 4 | 1042 | 72 | |
| BT056311.1 | - | 3 - 4 | 749 | 72 | ||
More Information
Set Membership: |
|

numb
Genomic Map