Assay Details
| Gene Symbol: Trx-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078802.4 | NP_523526.1 | 2 - 3 | 278 | 87 | |
| NM_164863.3 | NP_723475.1 | 3 - 4 | 469 | 87 | ||
| GenBank mRNA | AF220362.1 | - | 2 - 3 | 271 | 87 | |
| BT120324.1 | - | 2 - 3 | 275 | 87 | ||
More Information
Set Membership: |
|

Trx-2
Genomic Map