Assay Details
| Gene Symbol: CG42366 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273378.1 | NP_001260307.1 | 2 - 3 | 324 | 71 | |
| NM_164876.4 | NP_723501.2 | 2 - 3 | 324 | 71 | ||
More Information
Set Membership: |
|

CG42366
Genomic Map