Assay Details
| Gene Symbol: Cpr30F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164879.3 | NP_723504.1 | 2 | 125 | 64 | |
| GenBank mRNA | BT100145.3 | - | 1 | 54 | 64 | |
| KX531475.1 | - | 1 | 54 | 64 | ||
More Information
Set Membership: |
|

Cpr30F
Genomic Map