Assay Details
| Gene Symbol: me31B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078809.4 | NP_523533.2 | 2 - 3 | 346 | 115 | |
| NM_164898.2 | NP_723539.1 | 2 - 3 | 247 | 115 | ||
| GenBank mRNA | AY051663.1 | - | 2 - 3 | 275 | 115 | |
More Information
Set Membership: |
|

me31B
Genomic Map