Assay Details
| Gene Symbol: chico | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201843.2 | NP_001188772.1 | 2 - 3 | 444 | 105 | |
| NM_001273387.1 | NP_001260316.1 | 3 - 4 | 918 | 105 | ||
| NM_164899.3 | NP_723540.1 | 2 - 3 | 569 | 105 | ||
| GenBank mRNA | AF092046.1 | - | 2 - 3 | 519 | 105 | |
| AF154826.1 | - | 1 - 2 | 372 | 105 | ||
| BT016137.1 | - | 2 - 3 | 537 | 105 | ||
More Information
Set Membership: |
|

chico
Genomic Map