Assay Details
| Gene Symbol: chico | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201843.2 | NP_001188772.1 | 6 - 7 | 1201 | 83 | |
| NM_001273387.1 | NP_001260316.1 | 7 - 8 | 1675 | 83 | ||
| NM_164899.3 | NP_723540.1 | 6 - 7 | 1326 | 83 | ||
| GenBank mRNA | AF092046.1 | - | 6 - 7 | 1276 | 83 | |
| AF154826.1 | - | 5 - 6 | 83 | |||
| BT016137.1 | - | 6 - 7 | 1294 | 83 | ||
More Information
Set Membership: |
|

chico
Genomic Map