Assay Details
| Gene Symbol: Utx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273390.1 | NP_001260319.1 | 5 - 6 | 1060 | 120 | |
| NM_135524.4 | NP_609368.3 | 7 - 8 | 1332 | 120 | ||
| NM_164907.4 | NP_723550.2 | 5 - 6 | 867 | 120 | ||
| NM_164908.4 | NP_723551.2 | 5 - 6 | 855 | 120 | ||
| GenBank mRNA | AY129437.1 | - | 7 - 8 | 1397 | 120 | |
| BT010089.1 | - | 7 - 8 | 1332 | 120 | ||
More Information
Set Membership: |
|

Utx
Genomic Map