Assay Details
| Gene Symbol: Myo31DF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201855.2 | NP_001188784.1 | 2 - 3 | 417 | 77 | |
| NM_078814.4 | NP_523538.1 | 2 - 3 | 276 | 77 | ||
| NM_164933.3 | NP_723596.1 | 1 - 2 | 853 | 77 | ||
| GenBank mRNA | AY075567.1 | - | 1 - 2 | 832 | 77 | |
| U07595.1 | - | 2 - 3 | 290 | 77 | ||
More Information
Set Membership: |
|

Myo31DF
Genomic Map