Assay Details
| Gene Symbol: Myo31DF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201855.2 | NP_001188784.1 | 6 - 7 | 1551 | 92 | |
| NM_078814.4 | NP_523538.1 | 6 - 7 | 1410 | 92 | ||
| NM_164933.3 | NP_723596.1 | 5 - 6 | 1987 | 92 | ||
| GenBank mRNA | U07595.1 | - | 6 - 7 | 1424 | 92 | |
More Information
Set Membership: |
|

Myo31DF
Genomic Map