Assay Details
| Gene Symbol: Ge-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135642.3 | NP_609486.2 | 7 - 8 | 3189 | 84 | |
| NM_164957.2 | NP_723654.1 | 6 - 7 | 3272 | 84 | ||
| GenBank mRNA | BT004479.1 | - | 7 - 8 | 3169 | 84 | |
More Information
Set Membership: |
|

Ge-1
Genomic Map