Assay Details
| Gene Symbol: Ca-beta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259049.3 | NP_001245978.1 | 3 - 4 | 843 | 69 | |
| NM_001259050.3 | NP_001245979.2 | 3 - 4 | 1544 | 69 | ||
| NM_001273446.1 | NP_001260375.1 | 3 - 4 | 1544 | 69 | ||
| NM_001273447.1 | NP_001260376.1 | 5 - 6 | 905 | 69 | ||
| NM_001273448.2 | NP_001260377.1 | 3 - 4 | 1544 | 69 | ||
| NM_001298907.1 | NP_001285836.1 | 3 - 4 | 846 | 69 | ||
| NM_078822.4 | NP_523546.1 | 3 - 4 | 843 | 69 | ||
| NM_205962.3 | NP_995684.2 | 3 - 4 | 843 | 69 | ||
| NM_205963.4 | NP_995685.3 | 3 - 4 | 1544 | 69 | ||
| GenBank mRNA | U11074.1 | - | 3 - 4 | 441 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001273447.1 transcripts.
Set Membership: |
|

Ca-beta
Genomic Map