Assay Details
| Gene Symbol: Mal-B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201862.1 | NP_001188791.2 | 4 - 5 | 1733 | 62 | |
| NM_135679.5 | NP_609523.2 | 5 - 6 | 1741 | 62 | ||
| NM_164975.3 | NP_723693.3 | 4 - 5 | 1733 | 62 | ||
| GenBank mRNA | BT132976.1 | - | 5 - 6 | 1737 | 62 | |
More Information
Set Membership: |
|

Mal-B2
Genomic Map