Assay Details
| Gene Symbol: Fnta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273119.1 | NP_001260048.1 | 1 - 2 | 227 | 58 | |
| NM_135018.3 | NP_608862.1 | 1 - 2 | 227 | 58 | ||
| GenBank mRNA | AY118534.1 | - | 1 - 2 | 204 | 58 | |
More Information
Set Membership: |
|

Fnta
Genomic Map