Assay Details
| Gene Symbol: Scox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135040.4 | NP_608884.1 | 1 - 2 | 499 | 73 | |
| GenBank mRNA | AY089369.1 | - | 1 - 2 | 423 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scox| Gene Symbol: Scox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135040.4 | NP_608884.1 | 1 - 2 | 499 | 73 | |
| GenBank mRNA | AY089369.1 | - | 1 - 2 | 423 | 73 | |
Set Membership: |
|