Assay Details
| Gene Symbol: Rpn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135061.4 | NP_608905.1 | 2 - 3 | 934 | 62 | |
| GenBank mRNA | AF145313.1 | - | 1 - 2 | 764 | 62 | |
| AY070977.1 | - | 2 - 3 | 960 | 62 | ||
More Information
Set Membership: |
|

Rpn11
Genomic Map