Assay Details
| Gene Symbol: H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298698.1 | NP_001285627.1 | 1 - 2 | 558 | 59 | |
| NM_135082.3 | NP_608926.2 | 1 - 2 | 911 | 59 | ||
| GenBank mRNA | BT022140.1 | - | 1 - 2 | 885 | 59 | |
More Information
Set Membership: |
|

H15
Genomic Map