Assay Details
| Gene Symbol: mid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273167.2 | NP_001260096.1 | 1 - 2 | 463 | 57 | |
| NM_135083.4 | NP_608927.2 | 1 - 2 | 463 | 57 | ||
| GenBank mRNA | BT023769.1 | - | 1 - 2 | 449 | 57 | |
More Information
Set Membership: |
|

mid
Genomic Map