Assay Details
| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135090.4 | NP_608934.1 | 1 | 1487 | 132 | |
| GenBank mRNA | AY058443.1 | - | 1 | 1452 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq6| Gene Symbol: Coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135090.4 | NP_608934.1 | 1 | 1487 | 132 | |
| GenBank mRNA | AY058443.1 | - | 1 | 1452 | 132 | |
Set Membership: |
|