Assay Details
| Gene Symbol: CG17687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382040.1 | NP_001368937.1 | 2 - 3 | 408 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG17687| Gene Symbol: CG17687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001382040.1 | NP_001368937.1 | 2 - 3 | 408 | 97 | |
Set Membership: |
|