Assay Details
| Gene Symbol: Poc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274870.1 | NP_001261799.1 | 1 - 2 | 1010 | 79 | |
| NM_140383.2 | NP_648640.1 | 1 - 2 | 1010 | 79 | ||
| GenBank mRNA | AY069349.1 | - | 1 - 2 | 992 | 79 | |
More Information
Set Membership: |
|

Poc1
Genomic Map