Assay Details
| Gene Symbol: CG10738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259813.1 | NP_001246742.1 | 12 - 13 | 3033 | 71 | |
| NM_001259814.1 | NP_001246743.1 | 11 - 12 | 3027 | 71 | ||
| NM_140396.2 | NP_648653.2 | 11 - 12 | 3066 | 71 | ||
| NM_168547.2 | NP_729905.2 | 12 - 13 | 3072 | 71 | ||
| GenBank mRNA | BT099705.1 | - | 12 - 13 | 3033 | 71 | |
More Information
Set Membership: |
|

CG10738
Genomic Map