Assay Details
| Gene Symbol: CG10738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259814.1 | NP_001246743.1 | 7 - 8 | 2466 | 114 | |
| NM_140396.2 | NP_648653.2 | 7 - 8 | 2466 | 114 | ||
More Information
Set Membership: |
|

CG10738
Genomic Map