Assay Details
| Gene Symbol: CG10738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140396.2 | NP_648653.2 | 9 - 10 | 2742 | 55 | |
| NM_168547.2 | NP_729905.2 | 10 - 11 | 2748 | 55 | ||
More Information
Set Membership: |
|

CG10738
Genomic Map