Assay Details
| Gene Symbol: CG17364 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274887.1 | NP_001261816.1 | 4 - 5 | 1930 | 55 | |
| NM_001274888.1 | NP_001261817.1 | 4 - 5 | 1783 | 55 | ||
| NM_140417.3 | NP_648674.3 | 4 - 5 | 1783 | 55 | ||
| NM_176328.3 | NP_788506.1 | 4 - 5 | 1783 | 55 | ||
More Information
Set Membership: |
|

CG17364
Genomic Map