Assay Details
| Gene Symbol: Nprl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274889.1 | NP_001261818.1 | 2 - 3 | 175 | 79 | |
| NM_140423.3 | NP_648680.2 | 1 - 2 | 329 | 79 | ||
| NM_168570.1 | NP_729945.1 | 2 - 3 | 246 | 79 | ||
| GenBank mRNA | BT050578.1 | - | 1 - 2 | 329 | 79 | |
More Information
Set Membership: |
|

Nprl3
Genomic Map