Assay Details
| Gene Symbol: Sox21a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274898.1 | NP_001261827.1 | 2 - 3 | 932 | 62 | |
| NM_140437.2 | NP_648694.1 | 2 - 3 | 932 | 62 | ||
| GenBank mRNA | BT022137.3 | - | 1494 | 62 | ||
More Information
Set Membership: |
|

Sox21a
Genomic Map