Assay Details
| Gene Symbol: nan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274904.1 | NP_001261833.1 | 3 - 4 | 1125 | 96 | |
| NM_140439.3 | NP_648696.2 | 3 - 4 | 1125 | 96 | ||
| GenBank mRNA | AY262004.1 | - | 3 - 4 | 1126 | 96 | |
More Information
Set Membership: |
|

nan
Genomic Map