Assay Details
| Gene Symbol: nan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274904.1 | NP_001261833.1 | 4 - 5 | 1642 | 74 | |
| NM_140439.3 | NP_648696.2 | 4 - 5 | 1642 | 74 | ||
| GenBank mRNA | AY262004.1 | - | 4 - 5 | 1643 | 74 | |
More Information
Set Membership: |
|

nan
Genomic Map