Assay Details
| Gene Symbol: Tdrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140467.3 | NP_648724.1 | 2 - 3 | 177 | 65 | |
| GenBank mRNA | AY069766.1 | - | 2 - 3 | 175 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdrd3| Gene Symbol: Tdrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140467.3 | NP_648724.1 | 2 - 3 | 177 | 65 | |
| GenBank mRNA | AY069766.1 | - | 2 - 3 | 175 | 65 | |
Set Membership: |
|